Läs mer
Eurofins DiscoverX Fyn Protein, Active
Beschreibung
p59fyn is a member of the src-related gene family of non-receptor tyrosine kinases. Recent studies indicate that fyn kinase plays a critical role in mediating signal transduction through the T-cell receptor thereby initiating a signal transduction cascade leading to lymphokine secretion and cell proliferation. One of the earliest biochemical responses following T-cell receptor activation is an increase in tyrosine kinase activity and concomitant phosphorylation of several intracellular substrates. P59fyn kinase is an important mediator of this phosphorylation since it is physically associated with the T-cell receptor. Additional evidence supports the view that fyn participates in signal transduction pathways leading from multiple T-cell membrane proteins and hence may serve to integrate stimuli received from several independent receptor structures. In the case of the B-cell receptor, fyn is one of several kinases associated with the receptor and is activated following receptor cross-linking.
EMBL M14333
Spezifikation
Spezifikation
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 5% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvikt (g/mol) | 61.8 kDa |
| Kvantitet | 10 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Uttryckssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Full-length |
| Renhet eller kvalitetsklass | ≥56% |
| Protein | Fyn, active |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.