missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FYN Recombinant Human Protein
FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues.
Brand: Invitrogen™ PR4163D
Additional Details : Weight : 0.01000kg
Description
Fyn functions in a large number of pathways and has been shown to play a role in memory.
Specifications
NP_694592 | |
20mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
FYN | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
62.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |