missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ FZD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578242
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Positive Control: Synthetic peptide: 1:8000 Predicted Reactivity: Rat (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development.
Specifikationer
| FZD3 | |
| Polyclonal | |
| Unconjugated | |
| FZD3 | |
| AU020229; D930050A07Rik; frizzled 3; frizzled 3, seven transmembrane spanning receptor; frizzled class receptor 3; frizzled family receptor 3; frizzled homolog 3; frizzled homolog 3 (Drosophila); frizzled-3; Fz3; Fz-3; FZD3; hFz3; mFz3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14365, 7976 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.86 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q61086, Q9NPG1 | |
| FZD3 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human Frizzled 3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering