Läs mer
Tocris Bioscience™ G-15
Beskrivning
G-15 is a high affinity and selective GPER receptor antagonist (Ki = 20 nM) that displays no affinity for ERα and ERβ at concentrations up to 10 μM. Inhibits agonist-induced calcium mobilization in vitro (EC50 of ~185 nM) and antagonizes the antidepressive and renoprotective effects of estrogen in vivo.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (3aS*,4R*,9bR*)-4-(6-Bromo-1,3-benzodioxol-5-yl)-3a,4,5,9b-3H-cyclopenta[c]quinoline |
| CAS | 1161002-05-6 |
| Kvantitet | 10 mg |
| Mål | Estrogen and Related Receptor Antagonists |
| Molekylformel | C19H16BrNO2 |
| Renhet | 99% |