Läs mer
G 7460, TRC
Produktkod. 30483541
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483541 10 mg
30477601 100 mg
30484062 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483541 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM78314010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 102394-28-5
Molekylformel C14 H15 N5 O7 S
Molekylvikt (g/mol) 397.36
Synonym Benzoic acid, 4-hydroxy-2-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]-, methyl ester, G 7460
IUPAC-namn methyl 4-hydroxy-2-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoate
LEDER COC(=O)c1ccc(O)cc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2
Specifikationer
Kemiskt namn eller material Metsulfuron-methyl-4-hydroxy
Kvantitet 10 mg
InChI formel InChI=1S/C14H15N5O7S/c1-7-15-12(18-14(16-7)26-3)17-13(22)19-27(23,24)10-6-8(20)4-5-9(10)11(21)25-2/h4-6,20H,1-3H3,(H2,15,16,17,18,19,22)
Formel vikt 397.0692