missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ G6B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567267
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: DRVNLSCGGVS HPIRWVWAPS FPACKGLSKG RRPILWASSS GTPTVPPLQP FVGR Highest antigen sequence identity to the following orthologs - mouse 82%, rat 85%.
G6B is a member of the immunoglobulin (Ig) superfamily with 241 amino acids and is a glycosylated, plasma membrane-bound cell surface receptor, but soluble isoforms encoded by some transcript variants have been found in the endoplasmic reticulum and Golgi before being secreted. Multiple transcript variants encoding different isoforms have been reported. RT-PCR, Western blot, and flow cytometric analyses suggest that G6B is usually expressed on platelets and cross linking of G6B inhibits platelet aggregation and activation in a calcium-independent manner by agonists such as ADP and collagen-related peptide. G6B is an inhibitory surface receptor on platelets and may be an antithrombotic drug target. Isoform B is a putative inhibitory receptor. Isoform A may be its activating counterpart. It is known to bind to heparin. G6B is usually expressed in a restricted set of hematopoietic cell lines like K562, Molt4 and Jurkat and not detected in the cell lines like U937, Raji, Tk, HeLa, NKL, NK62 and YT.
Specifikationer
| G6B | |
| Polyclonal | |
| Unconjugated | |
| Mpig6b | |
| AU023871; C6orf25; chromosome 6 open reading frame 25; DADB-110M10.2; G6b; G6B-B; immunoglobulin receptor; immunoreceptor tyrosine-based inhibitory motif (ITIM) containing platelet receptor; Immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing platelet receptor; ITIM-receptor G6b-B; Megakaryocyte and platelet inhibitory receptor G6b; MPIG6B; NG31; Protein G6b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80739 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95866 | |
| Mpig6b | |
| Recombinant Human G6B. Recombinant protein control fragment (Product #RP-107903). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering