missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabapentin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Gabapentin is a water-soluble GABA analog with anticonvulsant and analgesic activities
• The exact mechanism of action is unknown but it has been described that this compound inhibits excitatory neuron activity by interacting with voltage-gated calcium channels in cortical neurons
Application
• Gabapentin has been used as the α2δ-ligand to examine whether the allyl isothiocyanate (AITC) sensitized thermal aversion assay might be useful as a screening method for the identification of presumptive analgesics
Specifications
Melting Point | 166.0°C |
Infrared Spectrum | Authentic |
Linear Formula | COOHCH2C6H10CH2NH2 |
Formula Weight | 171.24 |
Quantity | 25 g |
Chemical Name or Material | Gabapentin |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'