Läs mer
Gabapentin Related Compound B, TRC
Produktkod. 30135521
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135521 25 mg
30133544 100 mg
30010359 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135521 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG11727825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 133481-09-1
Molekylformel C9 H13 N O2
Molekylvikt (g/mol) 167.2
Synonym 1-Cyano-cyclohexaneacetic acid, (1-Cyanocyclohexyl)acetic acid, 1-Cyanocyclohexaneacetic acid
IUPAC-namn 2-(1-cyanocyclohexyl)acetic acid
LEDER OC(=O)CC1(CCCCC1)C#N
Specifikationer
Kemiskt namn eller material (1-cyanocyclohexyl)acetic acid
Anteckningar om renhetsgrad GC
Kvantitet 25 mg
InChI formel InChI=1S/C9H13NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-6H2,(H,11,12)
Formel vikt 167.0946
Procent renhet >95%