Learn More
Gabapentin Related Compound E, TRC
Product Code. 30136520
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136520 100 mg 100mg
17974625 10 mg 10mg
17930485 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136520 Supplier Toronto Research Chemicals Supplier No. TRCG117290100MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67950-95-2
Molecular Formula C9 H14 O4
Molecular Weight (g/mol) 186.21
Synonym 1-(Carboxymethyl)cyclohexanecarboxylic Acid, Gabapentin Imp. E (EP)
IUPAC Name 1-(carboxymethyl)cyclohexane-1-carboxylic acid
SMILES OC(=O)CC1(CCCCC1)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C9H14O4/c10-7(11)6-9(8(12)13)4-2-1-3-5-9/h1-6H2,(H,10,11)(H,12,13)
Formula Weight 186.0892
Percent Purity >95%
Chemical Name or Material 1-(Carboxymethyl)cyclohexanecarboxylic Acid
Grade HPLC