missing translation for 'onlineSavingsMsg'
Learn More

GABARAPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18724-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828604

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to GABARAPL2. It has no cross-reaction to other GABARAP protein.

Involved in intra-Golgi traffic. Modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1.
Specifications

Specifications

GABARAPL2
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P60520, P60521, P60522
ATG8, FLC3A, GABARAPL2, GATE 16, GATE16, GEF 2, GEF2
Rabbit
IgG
150 μL
-20°C
Primary
11345, 64670, 93739
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GABARAPL2
Liquid
Gabarapl2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers