Learn More
GAD1 Mouse anti-Human, Unconjugated, Clone: 2D5B7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67648-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840396
  • 1656.73 SEK / 20µL
Estimated Shipment: 30-04-2024
to see stock.
Description
Description
The GAD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GAD1. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The GAD1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
GAD1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Western Blot, Immunohistochemistry, ELISA
2D5B7
Unconjugated
20 μL
2571
IgG2a