missing translation for 'onlineSavingsMsg'
Learn More

GAGE2D Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12532-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16850284

  • 1656.73 SEK / 20µL
Estimated Shipment: 03-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GAGE2D, also named as GAGE8 and CT4.8, is expressed in testis and tumor tissues.
Specifications

Specifications

GAGE2D
Polyclonal
Unconjugated
GAGE2D
Cancer/testis antigen 4.8, CT4.8, G antigen 2D, G antigen 8, GAGE 8, GAGE2D, GAGE8
Rabbit
Antigen Affinity Chromatography
RUO
729408
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
0.57 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UEU5
GAGE2D
GAGE2D Fusion Protein Ag3230
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers