missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27552-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
GAL4 is a transcriptional activator which activates genes necessary for galactose catabolism. The DNA binding domain of GAL4 is contained in the first 74 amino acids and contains a "zinc finger" sequence motif. However, amino acids 1-147 have been shown to bind to DNA in vitro. While GAL4 (1-74) bind DNA specifically, it is with a lower affinity than the GAL4 (1-147). GAL4 (1-147) binds to a specific 17 base pair nucleotide sequence, but alone is insufficient to activate transcription. Either of the two acidic activating regions, activating region I (amino acids 148-238) or activating region II (amino acids 768-881), must be joined to activate transcription.Specifications
Galectin-4 | |
Polyclonal | |
Unconjugated | |
LGALS4 | |
Antigen NY CO 27, Gal 4, GAL4, Galectin-4, L 36 lactose binding protein, L36LBP, Lactose binding lectin 4, LGALS4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16855, 3960 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P56470, Q8K419 | |
Lgals4 | |
Galectin-4 Fusion Protein Ag26130 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |