missing translation for 'onlineSavingsMsg'
Learn More

Galectin 9 Mouse anti-Human, eFluor® 660, Clone: 9M1-3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9116-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15580077

  • 1160.00 SEK / 25 tests


Estimated Shipment
08-01-2021
Add to basket

Description

Description

Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin′s disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

Galectin 9
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O00182
HUAT, LGALS9A, ecalectin, gal-9, galectin 9, galectin-9, tumor antigen HOM-HD-21, urate transporter/channel protein
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
9M1-3
eFluor™ 660
LGALS9
Liquid
LGALS9
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3965
Videos
SDS
Documents

For Research Use Only.