missing translation for 'onlineSavingsMsg'
Learn More

Galidesivir hydrochloride, MedChemExpress

Product Code. 16481739
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
16481739 1 mg -
16410038 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481739 Supplier Medchem Express Supplier No. HY18649

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Galidesivir (BCX4430) hydrochloride, an adenosine analog and a direct-acting antiviral agent, disrupts viral RNA-dependent RNA polymerase (RdRp) activity.

Galidesivir hydrochloride is active in vitro against many RNA viral pathogens, including the filoviruses and emerging infectious agents such as MERS-CoV, SARS-CoV, and SARS-CoV-2. Galidesivir hydrochloride inhibits some negative-sense RNA viruses with EC50s ranging from ∼3 to ∼68 μM.

Specifications

Kemiskt namn eller material Galidesivir hydrochloride
Fysisk form Powder
CAS 222631-44-9
Färg Yellow
Kvantitet 1 mg
Anteckningar om renhetsgrad Research
Molekylformel C11H16ClN5O3
För användning med (applikation) COVID-19-anti-virus
Synonym BCX4430 hydrochlorideImmucillin-A hydrochloride
Löslighetsinformation DMSO : 105 mg/mL (347.99 mM; Need ultrasonic) ∣H2O : ≥ 41 mg/mL (135.88 mM)
LEDER O[C@H]1[C@H](C2=CNC3=C2N=CN=C3N)N[C@H](CO)[C@H]1O.Cl
Molekylvikt (g/mol) 301.73
Formel vikt 301.73
Procent renhet 98.51%
Kvalitet Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.