missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid, MP Biomedicals
Click to view available options
Quantity:
100 g
250 g
Specifications
Color | White |
Formula Weight | 170.1 |
Density | 1.694g/mL (Lit.) |
Quantity | 100 g |
Physical Form | Solid |
Chemical Name or Material | Gallic Acid |
Safety and Handling
Product Identifier
- Gallic Acid
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'