Läs mer
Invitrogen™ GALNT14 Monoclonal Antibody (2A8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522582
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide Sequence: KNGDDRQQWT KTGSHIEHIA SHLCLDTDMF GDGTENGKEI VVNPCESSLM SQHWDMVS.
GALNT14 (EC 2.4.1.41) belongs to a large subfamily of glycosyltransferases residing in the Golgi apparatus. GALNT enzymes catalyze the first step in the O-glycosylation of mammalian proteins by transferring N-acetyl-D-galactosamine (GalNAc) to peptide substrates.
Specifikationer
| GALNT14 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q96FL9 | |
| Galnt14 | |
| GALNT14 (NP_078848.2, 494 a.a. ~ 551 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2A8 | |
| Unconjugated | |
| Galnt14 | |
| 0610033M06Rik; FLJ12691; FLJ13977; GalNac-T10; GalNac-T14; GALNT14; GALNT15; Polypeptide GalNAc transferase 14; polypeptide N-acetylgalactosaminyltransferase 14; polypeptide N-acetylgalactosaminyltransferase 15; pp-GaNTase 14; protein-UDP acetylgalactosaminyltransferase 14; si:ch211-147c1.1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14); UNQ2434/PRO4994 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79623 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |