missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13962-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes GalNAc transferase 7, a member of the GalNAc-transferase family. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. Unlike other family members, this enzyme shows exclusive specificity for partially GalNAc-glycosylated acceptor substrates and shows no activity with non-glycosylated peptides. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation.Specifications
GALNT7 | |
Polyclonal | |
Unconjugated | |
GALNT7 | |
GALNAC T7, GalNAcT7, GALNT7, pp GaNTase 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108150, 29750, 51809 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80VA0, Q86SF2, Q9R0C5 | |
GALNT7 | |
GALNT7 Fusion Protein Ag4917 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |