Läs mer
Gamma-Butyrobetaine Hydrochloride, TRC
Produktkod. 30138375
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138375 10 g
30003774 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138375 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB75950010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6249-56-5
Molekylformel C7 H16 N O2 . Cl
Molekylvikt (g/mol) 181.66
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-, chloride (1:1), (3-Carboxypropyl)trimethylammonium chloride (6CI,7CI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-, chloride (9CI), Ammonium, (3-carboxypropyl)trimethyl-, chloride (8CI), N-Trimethyl-γ-aminobutyric acid chloride, γ-Butyrobetaine hydrochloride, (3-Carboxypropyl)trimethylammonium Chloride
IUPAC-namn 3-carboxypropyl(trimethyl)azanium;chloride
LEDER [Cl-].C[N+](C)(C)CCCC(=O)O
Specifikationer
Kemiskt namn eller material gamma-Butyrobetaine Hydrochloride
Kvantitet 10 g
InChI formel InChI=1S/C7H15NO2.ClH/c1-8(2,3)6-4-5-7(9)10;/h4-6H2,1-3H3;1H
Formel vikt 181.087