Läs mer
Gamma-Carboxyglutamic Acid Gamma,Gamma-Di-t-butyl Ester, TRC
Produktkod. 30361476
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30361476 5 mg
1 options
Produktkod. 30361476 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1784505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material gamma-Carboxyglutamic Acid gamma,gamma-Di-t-butyl Ester
CAS 56877-44-2
Molekylformel C14H25NO6
Kvantitet 5 mg
Synonym 3-Amino-1,1,3-propanetricarboxylic Acid 1,1-Bis(1,1-dimethylethyl) Ester
InChI formel InChI=1S/C14H25NO6/c1-13(2,3)20-11(18)8(7-9(15)10(16)17)12(19)21-14(4,5)6/h8-9H,7,15H2,1-6H3,(H,16,17)
LEDER CC(C)(C)OC(=O)C(CC(N)C(=O)O)C(=O)OC(C)(C)C
IUPAC-namn 2-amino-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonyl]-5-oxopentanoic acid
Molekylvikt (g/mol) 303.35
Formel vikt 303.1682