Learn More
Keystone Cleanroom Products Gamma-Irradiated 2 mil Polyethylene Trash Bag
Product Code. 17796869 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Includes:
1 Pack
10 Packs
2 Packs
5 Packs
Unit Size:
Pack of 10
Pack of 100
Pack of 40
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17796869 1 Pack Pack of 100
17706879 10 Packs Pack of 10
17726879 5 Packs Pack of 40
17716879 2 Packs Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17796869 Supplier Keystone Cleanroom Products Supplier No. S2428CP1
  • ONLINE EXCLUSIVE PRICE
    7860.00 SEK
    6090.05 SEK / Paket med 100
    Save 1769.95 SEK
    22% Av
Beräknad leverans: 11-06-2027
Simplify the removal of waste from your aseptic cleanroom without introducing external sources of microbial contamination, by using gamma irradiated trash liners.
  • Packaged in an ISO6 cleanroom
  • 2 mil clear polyethylene
  • Each package includes a lot number for traceability
  • Certificates of Conformance and Irradiation sent with shipment
  • Packaged in Tyvek™/Mylar Chevron Peel Pouches
Specifications
Autoclavable Not Autoclavable
Closure Type None
Color Clear
Includes 1 Pack
Material LDPE
Thickness (English) 2 mil
Dimensions (L x W) 71 x 61 cm (28 x 24 in.)
Product Type Cleanroom Bags
Capacity (Metric) 16.3 L
Thickness (Metric) 0.05 mm