Learn More
Gamma-Lindane-13C6, TRC
Product Code. 30474294
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
30474294 5 mg 5mg
1 options
Product Code. 30474294 Supplier Toronto Research Chemicals Supplier No. TRCL4659925MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material gamma-HCH-13C6
CAS 104215-85-2
Molecular Formula 13C6 H6 Cl6
Quantity 5 mg
Synonym (1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane-13C6, gamma-HCH 13C6
InChI Formula InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+/i1+1,2+1,3+1,4+1,5+1,6+1
SMILES Cl[13C@@H]1[13C@@H](Cl)[13C@@H](Cl)[13C@H](Cl)[13C@@H](Cl)[13C@H]1Cl
IUPAC Name 1,2,3,4,5,6-hexachloro(1,2,3,4,5,6-^{13}C_{6})cyclohexane
Molecular Weight (g/mol) 296.79
Formula Weight 293.8802