Läs mer
Gamma-Lumi (-)-Colchicine, TRC
Produktkod. 30012813
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012813 25 mg
30009620 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012813 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47431025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6901-14-0
Molekylformel C22H25NO6
Molekylvikt (g/mol) 399.44
Synonym Acetamide, N-[(7S,7bS,10aR)-5,6,7,7b,8,10a-hexahydro-1,2,3,9-tetramethoxy-8-oxobenzo[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl]-, Acetamide, N-(5,6,7,7b,8,10a-hexahydro-1,2,3,9-tetramethoxy-8-oxobenzo[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl)-, [7S-(7α,7bα,10aα)]-, γ-Lumicolchicine (6CI,7CI,8CI), N-[(7S,7bS,10aR)-5,6,7,7b,8,10a-Hexahydro-1,2,3,9-tetramethoxy-8-oxobenzo[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl]acetamide
LEDER COC1=C[C@@H]2[C@H](C1=O)C3=C2c4c(CC[C@@H]3NC(=O)C)cc(OC)c(OC)c4OC
Specifikationer
Kemiskt namn eller material gamma-Lumicolchicine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C22H25NO6/c1-10(24)23-13-7-6-11-8-15(27-3)21(28-4)22(29-5)16(11)17-12-9-14(26-2)20(25)18(12)19(13)17/h8-9,12-13,18H,6-7H2,1-5H3,(H,23,24)/t12-,13+,18-/m1/s1
Formel vikt 399.1682
Procent renhet >95%