Läs mer
gamma-Methylaminobutyraldehyde Diethyl Acetal, TRC
Produktkod. 30472600
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30472600 100 mg
1 options
Produktkod. 30472600 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM285740100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material gamma-Methylaminobutyraldehyde','Diethyl Acetal
CAS 114094-45-0
Molekylformel C9 H21 N O2
Kvantitet 100 mg
Synonym Zolmitriptan Impurity 17
InChI formel InChI=1S/C9H21NO2/c1-4-11-9(12-5-2)7-6-8-10-3/h9-10H,4-8H2,1-3H3
LEDER CCOC(CCCNC)OCC
IUPAC-namn 4,4-diethoxy-N-methylbutan-1-amine
Molekylvikt (g/mol) 175.27
Formel vikt 175.1572