Läs mer
gamma-Secretase Modulators, MedChemExpress
Produktkod. 16428138
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16428138 10 mg -
16400598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16428138 Leverantör Medchem Express Leverantörsnummer HY50900
MedChemExpress gamma-Secretase Modulators (Amyloid-β production inhibitor) is a Amyloid-β production inhibitor.
gamma-Secretase Modulators (Amyloid-β production inhibitor) is a Amyloid-β production inhibitor.
Specifikationer
Kemiskt namn eller material gamma-Secretase Modulators
CAS 937812-80-1
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C26H24F3N3O3
Kvantitet 10 mg
Synonym Amyloid-β production inhibitor γ-Secretase Modulators
Löslighetsinformation DMSO : ≥ 100 mg/mL (206.83 mM)
LEDER O=C1N2[C@@H](COC[C@]2([H])CC/C1=C\C3=CC(OC)=C(C=C3)N4C=NC(C)=C4)C5=CC(F)=C(F)C(F)=C5
Molekylvikt (g/mol) 483.48
Formel vikt 483.48
Procent renhet 98.07%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neurodegeneration