Läs mer
Invitrogen™ gamma Synuclein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519746
Beskrivning
PA5-19746 targets gamma Synuclein in WB applications and shows reactivity with Human samples. The PA5-19746 immunogen is synthetic peptide conjugated to KLH derived from within residues 100 to the C-terminus of Human gamma Synuclein. PA5-19746 detects gamma Synuclein which has a predicted molecular weight of approximately 15 kDa.
This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
Specifications
| gamma Synuclein | |
| Polyclonal | |
| Unconjugated | |
| SNCG | |
| BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6623 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.9 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| O76070 | |
| SNCG | |
| Synthetic peptide conjugated to KLH derived from within residues 100 to the C-terminus of Human gamma Synuclein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.