Läs mer
Gamma-Valprolactone, TRC
Produktkod. 30451834
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30451834 250 mg
1 options
Produktkod. 30451834 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV094800250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material gamma-Valprolactone
CAS 40923-58-8
Molekylformel C8 H14 O2
Kvantitet 250 mg
Synonym 2(3H)-Furanone','dihydro-5-methyl-3-propyl-, Valeric acid','4-hydroxy-2-propyl-','lactone (6CI), Valeric acid','γ-hydroxy-α-propyl-','lactone (4CI), Dihydro-5-methyl-3-propyl-2(3H)-furanone, γ-Valprolactone, 5-Methyl-3-propyloxolan-2-one
InChI formel InChI=1S/C8H14O2/c1-3-4-7-5-6(2)10-8(7)9/h6-7H,3-5H2,1-2H3
LEDER CCCC1CC(C)OC1=O
IUPAC-namn 5-methyl-3-propyloxolan-2-one
Molekylvikt (g/mol) 142.2
Formel vikt 142.0994