Läs mer
Tocris Bioscience™ Ganaxolone
Beskrivning
Ganaxolone is a potent positive allosteric modulator of GABAA receptors. Enhances GABA-evoked chloride currents in Xenopus oocytes expressing GABAA receptors (EC50 values are 94, 122 and 213 nM for α2β1γ2L, α3β1γ2L and α1β1γ2L receptors respectively). Exerts anticonvulsive effects in a broad range of animal seizure models.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (3α,5α)-3-Hydroxy-3-methyl-pregnan-20-one |
| CAS | 38398-32-2 |
| Kvantitet | 10 mg |
| Mål | GABA-A Receptor Modulators |
| Molekylformel | C22H36O2 |