Läs mer
Ganciclovir EP Impurity J, Mikromol™
Produktkod. 30051411
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051411 100 mg
30050324 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051411 Leverantör Mikromol Leverantörsnummer MM0485.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ganciclovir EP Impurity J
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[2-(Propanoylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy]propane-1,3-diyl dipropanoate (Ganciclovir Tripropionate)
CAS 177216-32-9
Analyt- eller komponentnamn 2-[2-(Propanoylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy]propane-1,3-diyl dipropanoate (Ganciclovir Tripropionate)
Frakt skick Room Temperature
Molekylformel C18 H25 N5 O7
Certifikat och efterlevnad ISO 17025
Synonym 2-[2-(Propanoylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy]propane-1,3-diyl dipropanoate, Ganciclovir Imp. J (EP), Ganciclovir Tripropionate, Ganciclovir Impurity J
InChI formel InChI=1S/C18H25N5O7/c1-4-12(24)20-18-21-16-15(17(27)22-18)19-9-23(16)10-30-11(7-28-13(25)5-2)8-29-14(26)6-3/h9,11H,4-8,10H2,1-3H3,(H2,20,21,22,24,27)
LEDER CCC(=O)NC1=Nc2c(ncn2COC(COC(=O)CC)COC(=O)CC)C(=O)N1
IUPAC-namn [2-[[6-oxo-2-(propanoylamino)-1H-purin-9-yl]methoxy]-3-propanoyloxypropyl] propanoate
Molekylvikt (g/mol) 423.42
Kvantitet 100 mg
Formel vikt 423.1754 g/mol