Läs mer
Ganciclovir Mono-O-propionate, TRC
Produktkod. 17920725
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17920725 50 mg
17992285 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17920725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG23502050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 194159-18-7
Molekylformel C12 H17 N5 O5
Molekylvikt (g/mol) 311.29
Synonym (2RS)-2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]-3-hydroxy-propyl Propionate, Ganciclovir Monopropionate, Ganciclovir Mono-O-propionate, Ganciclovir Imp. B (EP), 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[[1-(hydroxymethyl)-2-(1-oxopropoxy)ethoxy]methyl]-, 2-Amino-1,9-dihydro-9-[[1-(hydroxymethyl)-2-(1-oxopropoxy)ethoxy]methyl]-6H-purin-6-one, Valganciclovir Hydrochloride Imp. J (EP)
IUPAC-namn [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-3-hydroxypropyl] propanoate
LEDER CCC(=O)OCC(CO)OCn1cnc2C(=O)NC(=Nc12)N
Specifikationer
Kemiskt namn eller material Ganciclovir Mono-O-propionate
Kvantitet 50 mg
InChI formel InChI=1S/C12H17N5O5/c1-2-8(19)21-4-7(3-18)22-6-17-5-14-9-10(17)15-12(13)16-11(9)20/h5,7,18H,2-4,6H2,1H3,(H3,13,15,16,20)
Formel vikt 311.123