Learn More
Ganciclovir Triacetate, Mikromol™
Product Code. 18091710
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
18091710 25 mg 25mg
1 options
Product Code. 18091710 Supplier Mikromol Supplier No. MM0485.140025
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ganciclovir Triacetate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ganciclovir Triacetate
CAS 86357-14-4
Analyte or Component Names Ganciclovir Triacetate
Shipping Condition Room Temperature
Molecular Formula C15 H19 N5 O7
Certifications/Compliance ISO 17025
Synonym N-[9-[[2-(Acetyloxy)-1-[(acetyloxy)methyl]ethoxy]methyl]-6,9-dihydro-6-oxo-1H-purin-2-yl]acetamide, N-Acetyl-di-O-acetyl Ganciclovir, Ganciclovir Triacetate, Triacetyl Ganciclovir
InChI Formula InChI=1S/C15H19N5O7/c1-8(21)17-15-18-13-12(14(24)19-15)16-6-20(13)7-27-11(4-25-9(2)22)5-26-10(3)23/h6,11H,4-5,7H2,1-3H3,(H2,17,18,19,21,24)
SMILES CC(=O)NC1=Nc2c(ncn2COC(COC(=O)C)COC(=O)C)C(=O)N1
IUPAC Name [2-[(2-acetamido-6-oxo-1H-purin-9-yl)methoxy]-3-acetyloxypropyl] acetate
Molecular Weight (g/mol) 381.34
Quantity 25 mg
Formula Weight 381.1284 g/mol