Läs mer
STEMCELL Technologies GANT 58
Beskrivning
- GANT 58 inhibits the Hedgehog signaling pathway downstream of Smoothened and Suppressor of Fused (SUFU) leading to GLI1 nuclear accumulation (Joo et al.; Stanton & Peng)
- GANT 58 demonstrates antiproliferative and antitumor activity in vivo (Beauchamp et al.; Joo et al.)
- Reduces anchorage-independent growth in Ewing Sarcoma cells (Beauchamp et al.; Joo et al.)
- Inhibits prostate cancer tumor growth (Lauth et al.)
- Causes cell cycle arrest and apoptosis in acute leukemia T cells (Hou et al.)
Specifikationer
Specifikationer
| CAS | 64048-12-0 |
| Synonym | NSC 75503 |
| Kvantitet | 10 mg |
| Molekylformel | C24H16N4S |
| Renhet | ≥98% |