Läs mer
GAPDH Antibody (CL3266) - Azide and BSA Free, Novus Biologicals™
Produktkod. 30300010 Handla allt Bio Techne Produkter
Change view
Kvantitet:
100 μL
Produktkod. Kvantitet
30300010 100 μL
1 options
Produktkod. 30300010 Leverantör Novus Biologicals Leverantörsnummer NBP344492
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Mouse Monoclonal Antibody
GAPDH Monoclonal antibody specifically detects GAPDH in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry,KnockDown
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen GAPDH
Användningsområden Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, KnockDown
Klassificering Monoclonal
Klona CL3266
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:50000 - 1:100000
Formulering Lyophilized in 5% Trehalose.
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Värd art Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P04406, with the exact immunogen sequence remaining proprietary.
Reningsmetod Protein A purified
Kvantitet 100 μL
Regulatorisk status RUO
Forskningsdisciplin Alzheimers Research, Apoptosis, Autophagy, Cancer, Chromatin Research, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primär eller sekundär Primary
Gen-ID (Entrez) 2597
Rekonstitution Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Målarter Human
Innehåll och lagring Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
Form Purified
Isotyp IgG2a