missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GAPDH, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ LFP0008
Additional Details : Weight : 0.99000kg
Description
GAPDH (Hu) Recombinant Protein, LF-P0008, from Invitrogen.
GAPDH (Glyceraldehyde-3-phosphate dehydrogenase) is a catalytic enzyme commonly known to be involved in glycolysis. GAPDH exists as a tetramer of identical 37-kDa subunits and catalyzes the reversible reduction of 1,3-bisphosphoglycerate to glyceraldehyde 3-phosphophate in the presence of NADPH. Apart from playing a key role in glycolysis, GAPDH is ubiquitously expressed and displays other activities unrelated to its glycolytic function. GAPDH is reported to be involved in the processes of DNA replication, DNA repair, nuclear RNA export, membrane fusion and microtubule bundling. Studies provide evidence of GAPDH playing an essential part in gene expression observed in apoptosis and as part of the cellular phenotype of age-related neurodegenerative diseases. Further, GAPDH is involved in other cellular processes ranging from membrane fusion, and neuronal apoptosis in cancer. GAPDH is reported to bind to a variety of other proteins, including the amyloid precursor protein, mutations in which cause some forms of Alzheimer′s disease (AD), and the polyglutamine tracts of Huntingtin, the protein product aberrant forms of which are causative of Huntington′s disease. Associations between GAPDH, actin and tubulin have also be reported. Since GAPDH is expressed at high levels in most tissues, it is useful as protein loading control in Western Blot analysis. This protein corresponds to human GAPDH (1-335 aa), which has a molecular weight of 35.9 kDa. Specific activity: up to 30 U/mg. Purity: >95% by SDS-PAGE. After reconstitution, store at -80°C and avoid freeze and thaw cycles.Specifications
P04406 | |
0.02M HEPES with no preservative; pH 7.4 | |
Human GAPDH | |
0.5mg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Glyceraldehyde-3-Phosphate Dehydrogenase | |
GAPDH | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
Control | |
2597 | |
7.4 | |
E. coli | |
RUO | |
Human GAPDH | |
Human | |
Hu | |
Recombinant | |
Recombinant |