Learn More
GARNL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554590
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LVSTDAGVLL VDDDLPSVPV FDRTLPVKQM HVLETLDLLV LRADKGKDAR LFVFRLSALQ KGLEGKQAGK SRSDCRENKL EKTKGCHLYA INTHHS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
GARNL3 Polyclonal antibody specifically detects GARNL3 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GARNL3 | |
Polyclonal | |
Unconjugated | |
GARNL3 | |
AW120551; bA356B19.1; Garnl3; GTPase activating RANGAP domain-like 3; GTPase activating Rap/RanGAP domain like 3; GTPase activating Rap/RanGAP domain-like 3; GTPase-activating Rap/Ran-GAP domain-like protein 3; RGD1564019 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84253 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VVW2 | |
GARNL3 | |
Recombinant protein corresponding to Human GARNL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only