Läs mer
BOLA Gas Frits
Kontakter:
5 mm
7 mm
6 x 1 mm
Beskrivning
- Microporous PTFE with pore size 5 μm for steady sparkling of the flowing gas.
- Suitable for scrubber bottles and columns with M 8x1 thread and for gas inlet tubes.
Specifikationer
Specifikationer
| Autoklaveringsbar | Autoclavable |
| Kontakter | 5 mm |
| Disponibel | Reusable |
| Provvolym | 5 μL |
| Sterilitet | Non-sterile |
| Längd (metrisk) | 15 mm |