Learn More
3M™ Gas and Vapor Cartridge, 6000 Series
Combines excellent protection with superb comfort and balance. The 3M Gas and Vapor Cartridge is trapezoidal shaped and works with half- and full-facemasks that have bayonet connections systems.
Brand: 3M™ 7000034749
Additional Details : Weight : 0.76050kg
Description
Each filter has a colored band using the EN14387 classification system, to denote the nature of the protection it provides.
- Trapezoidal-shaped gas and vapor filters combine excellent protection with superb comfort and balance
- Lightweight, low breathing resistance, and well balanced when fitted to appropriate mask
- Provides excellent field of vision as a result of the unique trapezoidal shape.
- Filters can be used on 3M™ Facemasks fitted with a bayonet connections system