Learn More
3M™ Gas, Vapor, and Particulate Filter, 6000 Series
Provides excellent field of vision as a result of the unique trapezoidal shape. The 3M Gas, Vapor, and Particulate Filter uses bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks.
Brand: 3M™ 7000021315
Additional Details : Weight : 0.30600kg
Description
Unique, compatible Bayonet Filter Attachment System allows different types of filters to just be clicked into place ensuring optimum ease of use.- Lightweight, low breathing resistance, and well-balanced when fitted to mask
- Provides excellent field of vision as a result of the unique trapezoidal shape
- Bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks