Learn More
- Home
- Products
- Pumps
- Pump Parts and Accessories
- Pump Seals
- RETSCH Gasket For Grinding Jar
RETSCH Gasket For Grinding Jar
Product Code. 15340984 Shop All Retsch Products
Change view
Click to view available options
Material:
Stainless Steel
Stainless Steel, PTFE
Zirconium Oxide
Zirconium Oxide, Tungsten Carbide
For Use With (Equipment):
1.5 mL Grinding Jar
10 mL Grinding Jar
25 mL Grinding Jar
35 mL Grinding Jar
5 mL Grinding Jar
50 mL Grinding Jar
Quantity:
1 Pk.
10 Pk.
Unit Size:
Each
Pack of 10
13 product options available for selection
| Product Code. | Material | For Use With (Equipment) | Quantity | Unit Size |
|---|---|---|---|---|
| 15340984 | Zirconium Oxide | 25 mL Grinding Jar | 1 Pk. | Each |
| 11924362 | Stainless Steel, PTFE | 1.5 mL Grinding Jar | 10 Pk. | Each |
| 15350984 | Stainless Steel, PTFE | 10 mL Grinding Jar | 1 Pk. | Each |
| 10466739 | Stainless Steel, PTFE | 10 mL Grinding Jar | 10 Pk. | Pack of 10 |
| 15330984 | Stainless Steel, PTFE | 25 mL Grinding Jar | 1 Pk. | Each |
| 15320984 | Stainless Steel, PTFE | 35 mL Grinding Jar | 1 Pk. | Each |
| 10250315 | Zirconium Oxide, Tungsten Carbide | 25 mL Grinding Jar | 10 Pk. | Pack of 10 |
| 11944362 | Stainless Steel | 35 mL Grinding Jar | 10 Pk. | Each |
| 15310984 | Stainless Steel, PTFE | 50 mL Grinding Jar | 1 Pk. | Each |
| 10472080 | Zirconium Oxide | 35 mL Grinding Jar | 10 Pk. | Pack of 10 |
| 15300984 | Stainless Steel, PTFE | 5 mL Grinding Jar | 1 Pk. | Each |
| 11934362 | Stainless Steel, PTFE | 5 mL Grinding Jar | 10 Pk. | Each |
| 10290773 | Stainless Steel | 25 mL Grinding Jar | 10 Pk. | Pack of 10 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
Product Code.
15340984
Supplier
RETSCH
Supplier No.
03.111.0296
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Specifications
| For Use With (Equipment) | 25 mL Grinding Jar |
| Quantity | 1 Pk. |
| Material | Zirconium Oxide |
| Type | Gasket |