Läs mer
BOLA Gaskets for Caps
Produktkod. 31891578
Change view
Produkttyp:
Septa
Produktkod. Produkttyp
31891578 Septa
1 options
Produktkod. 31891578 Leverantör BOLA Leverantörsnummer H97345
  • ONLINE EXCLUSIVE PRICE
    611.20 SEK
    523.89 SEK / Paket med 20
    Save 87.31 SEK
    14% Av
Beräknad leverans: 20-11-2026
Designed specifically for use with BOLA screw caps with apertures, serving as seal or septum replacements
  • Upper side made of PTFE, lower side made of silicone-elastomer for balancing unevennesses on sealing surfaces
  • After assembly, the product is only exposed to PTFE
  • Abmessungen: For thread GL 45 Dia. of gasket 43.2 mm, Thickness of gasket 3.3 mm
Specifikationer
Färg White
Material PTFE-faced Silicone
Sterilitet Non-sterile
Autoklaveringsbar Not Autoclavable
Form Round
Produkttyp Septa