missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gastrin I, Rat

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66510.|D

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15437899

  • 1541.44 SEK / 0.50mg


Estimated Shipment
23-10-2020
Add to basket

Specifications

Specifications

2126.28
Rat
Solid
RUO
Gastrin I, rat
HPLC
0.5mg
Glp-Arg-Pro-Pro-Met-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2
-30° to -10°C
Videos
SDS Documents

For Research Use Only