missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Mouse anti-Human, Clone: 7G9C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60346-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
Gastrin is a 17 amino acid peptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Study of gastrin may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS.Specifications
Gastrin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01350 | |
GAST | |
Gastrin Fusion Protein Ag12795 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin) | |
7G9C3 | |
Unconjugated | |
GAST | |
G14 Gastrin 6, G17 Gastrin 14, G34 Gastrin, G52 Big gastrin, G6, GAS, GAST, gastrin, Gastrin 17, Gastrin 34, Gastrin component I Gastrin 52, Gastrin component II, Gastrin component III | |
Mouse | |
Protein A | |
RUO | |
2520 | |
-20°C | |
Liquid |