missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATAD2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GATAD2B |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824245
|
Proteintech
25679-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814245
|
Proteintech
25679-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a zinc finger protein transcriptional repressor. The encoded protein is part of the methyl-CpG-binding protein-1 complex, which represses gene expression by deacetylating methylated nucleosomes. Mutations in this gene are linked to intellectual disability and dysmorphic features associated with cognitive disability.Specifications
GATAD2B | |
Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q8WXI9 | |
57459 | |
GATAD2B Fusion Protein Ag22615 | |
Primary | |
-20°C | |
GATAD2B |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GATAD2B, KIAA1150, p66/p68, P66beta, RP11 216N14.6 | |
GATAD2B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |