missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GBP2 (Human) Recombinant Protein
Human GBP2 full-length ORF ( AAH22272, 1 a.a. - 481 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002634-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAPEINLPGPMSLIDNTKGQLVVNPEALKILSAITQPVVVVAIVGLYRTGKSYLMNKLAGKKNGFSLGSTVKSHTKGIWMWCVPHPKKPEHTLVLLDTEGLGDIEKGDNENDSWIFALAILLSSTFVYNSMGTINQQAMDQLHYVTELTDRIKANSSPGNNSVDDSADFVSFFPAFVWTLRDFTLELEVDGEPITADDYLELSLKLRKGTDKKSKSFNDPRLCIRKFFPKRKCFVFDWPAPKKYLAHLEQLKEEELNPDFIEQVAEFCSYILSHSNVKTLSGGIPVNGPRLESLVLTYVNAISSGDLPCMENAVLALAQIENSAAVEKAIAHYEQQMGQKVQLPTETLQELLDLHRDSEREAIEVFMKNSFKDVDQMFQRKLGAQLEARRDDFCKQNSKASSDCCMALLQDIFGPLEEDVKQGTFSKPGGYRLFTQKLQELKNKYYQVPRKGIQAKEVLKKYLESKEDVADALLQTDQSLS
Specifications
AAH22272 | |
guanylate binding protein 2, interferon-inducible | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2634 | |
Wheat germ expression system | |
25 μg | |
GBP2 | |
GST |