Läs mer
Invitrogen™ GBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588902
Beskrivning
Immunogen sequence: QDIFGPLEED VKQGTFSKPG GYRLFTQKLQ ELKNKYYQVP RKGIQAKEVL KKYLESKEDV ADALLQTDQS LSEKEKAIEV ERIKAESAEA AKKMLEEIQK KNEEMMEQKE K; Positive Samples: 293T, A375; Cellular Location: Cytoplasm, Golgi apparatus membrane, Lipid-anchor, Membrane, perinuclear region.
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
Specifikationer
| GBP2 | |
| Polyclonal | |
| Unconjugated | |
| GBP2 | |
| GBP2; GBP-2; GTP-binding protein 2; guanine nucleotide-binding protein 2; guanylate binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; mGBP2; mGBP-2; p67 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14469, 2634 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.41 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P32456, Q9Z0E6 | |
| GBP2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.