missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27299-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.Specifications
GBP2 | |
Polyclonal | |
Unconjugated | |
GBP2 | |
GBP 2, GBP2, GTP binding protein 2, HuGBP 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14469, 2634 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P32456, Q9Z0E6 | |
Gbp2 | |
GBP2 Fusion Protein Ag25433 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |