Läs mer
Tocris Bioscience™ GBR 12783 dihydrochloride
GREENER_CHOICE
Produktkod. 18045352
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18045352 50 mg
18034472 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18045352 Leverantör Tocris Bioscience™ Leverantörsnummer 0513/50
Potent and selective DA uptake inhibitor
A very potent and selective inhibitor of dopamine uptake (IC50 for inhibition of [3H]-dopamine uptake in rat striatal synaptosomes is 1.8 nM).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67469-75-4
Molekylformel C28H34Cl2N2O
Specifikationer
Kemiskt namn eller material GBR 12783 dihydrochloride
Kvantitet 50 mg
Formel vikt 485.5
Renhet >99%
Molekylvikt (g/mol) 485.493