Läs mer
GBR 12783 Dihydrochloride, TRC
Produktkod. 30403858
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30403858 1 mg
1 options
Produktkod. 30403858 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG2680131MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material GBR 12783 Dihydrochloride
CAS 67469-75-4
Molekylformel C28H32N2O • 2(HCl)
Kvantitet 1 mg
Synonym 1-(2-Diphenylmethoxyethyl)-4-(3-phenyl-2-propenyl)-piperazine Dihydrochloride, Lu 2-098, MW 486
InChI formel InChI=1S/C28H32N2O/c1-4-11-25(12-5-1)13-10-18-29-19-21-30(22-20-29)23-24-31-28(26-14-6-2-7-15-26)27-16-8-3-9-17-27/h1-17,28H,18-24H2/b13-10+
LEDER N1(CCN(CC1)C/C=C/C2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4
Molekylvikt (g/mol) 412.5723646
Formel vikt 412.2515