Läs mer
Tocris Bioscience™ GC 1
GREENER_CHOICE
Produktkod. 30001642
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001642 10 mg
30001660 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001642 Leverantör Tocris Bioscience™ Leverantörsnummer 4554/10
High affinity thyroid receptor α (TRα) and TRβ agonist; thyromimetic
Thyromimetic; high affinity agonist at thyroid hormone receptor (TR) β and TRα receptors (KD values are 67 and 440 pM respectively). Displays 5- and 100-fold greater potency than the endogenous agonist T3 in vitro at TRα1 and TRβ1 receptors respectively. Promotes reverse cholesterol transport in chow- and fat-fed mice. Induces proliferation of hepatocytes and pancreatic acinar cells in rats independent of prior tissue injury.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 211110-63-3
Molekylformel C20H24O4
Specifikationer
Kemiskt namn eller material GC 1
Kvantitet 10 mg
Formel vikt 328.4
Renhet >98%
Molekylvikt (g/mol) 328.408