missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDPD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | GDPD5 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836144
|
Proteintech
16494-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16826144
|
Proteintech
16494-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with maligt choline phospholipid metabolite profiles in human breast cancer.Specifications
GDPD5 | |
Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q640M6, Q8WTR4 | |
233552, 499211, 81544 | |
GDPD5 Fusion Protein Ag9622 | |
Primary | |
-20°C | |
GDPD5 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GDE2, GDPD5, PP1665, PP9363, UNQ1850/PRO3580 | |
Gdpd5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |